Assay Details
Gene Symbol: Nptn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019380.1 | NP_062253.1 | 2 - 3 | 581 | 74 | |
XM_006243172.2 | XP_006243234.1 | 2 - 3 | 594 | 74 | ||
GenBank mRNA | X99338.1 | - | 2 - 3 | 581 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
