Assay Details
Gene Symbol: Nptn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019380.1 | NP_062253.1 | 3 - 4 | 751 | 98 | |
XM_006243172.2 | XP_006243234.1 | 3 - 4 | 764 | 98 | ||
XM_006243174.2 | XP_006243236.1 | 2 - 3 | 425 | 98 | ||
XM_008766330.1 | XP_008764552.1 | 2 - 3 | 425 | 98 | ||
GenBank mRNA | BC070947.1 | - | 2 - 3 | 281 | 98 | |
X99337.1 | - | 2 - 3 | 454 | 98 | ||
X99338.1 | - | 3 - 4 | 751 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
