Assay Details
Gene Symbol: Nptn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019380.1 | NP_062253.1 | 5 - 6 | 974 | 63 | |
XM_006243172.2 | XP_006243234.1 | 5 - 6 | 987 | 63 | ||
XM_006243174.2 | XP_006243236.1 | 4 - 5 | 648 | 63 | ||
XM_008766330.1 | XP_008764552.1 | 4 - 5 | 648 | 63 | ||
GenBank mRNA | BC070947.1 | - | 4 - 5 | 504 | 63 | |
X99337.1 | - | 4 - 5 | 677 | 63 | ||
X99338.1 | - | 5 - 6 | 974 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
