Assay Details
Gene Symbol: Hacd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106831.1 | NP_001100301.1 | 9 - 10 | 1083 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
I-kappaB kinase/NF-kappaB signaling
JNK cascade
Rho protein signal transduction
fatty acid elongation
very long-chain fatty acid biosynthetic process
positive regulation of viral genome replication
positive regulation by virus of viral protein levels in host cell
3-hydroxyacyl-CoA dehydratase activity
enzyme binding
I-kappaB kinase/NF-kappaB signaling
JNK cascade
Rho protein signal transduction
fatty acid elongation
very long-chain fatty acid biosynthetic process
positive regulation of viral genome replication
positive regulation by virus of viral protein levels in host cell
3-hydroxyacyl-CoA dehydratase activity
enzyme binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P336608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13521 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444727 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17465 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P336608 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P336608 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P336608 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P516074 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 104020 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 56184 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10918260 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918263 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918261 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918259 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918264 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918262 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10918258 | - | - |
Rat Genome 230 2.0 Array | 1373469_PM_at | - | - |
Rat Genome 230 2.0 Array | 1383587_PM_at | - | - |
Rat Genome 230 2.0 Array | 1373469_at | - | - |
Rat Genome 230 2.0 Array | 1384539_at | - | - |
Rat Genome 230 2.0 Array | 1384539_PM_at | - | - |
Rat Genome 230 2.0 Array | 1383587_at | - | - |
Affymetrix HT Rat Focus Array | 1383587_at | - | - |
Affymetrix HT Rat Focus Array | 1383587_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373469_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373469_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0800022514.rn.1 | TC0800002772.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800022515.rn.1 | TC0800002772.rn.1 | RTA TC |
Rat Expression Set 230 | 1373469_PM_at | - | - |
Rat Expression Set 230 | 1373469_at | - | - |
Rat Expression Set 230 | 1383587_at | - | - |
Rat Expression Set 230 | 1383587_PM_at | - | - |
Rat Genome U34 Set | rc_AA891196_at | - | - |
Rat Genome U34 Set | rc_AA875148_at | - | - |
Rat Genome U34 Set | rc_AA946253_at | - | - |
Rat Genome U34 Set | rc_AI045163_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21796093 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350417 | - | - |
