Assay Details
Gene Symbol: Adpgk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100723.1 | NP_001094193.1 | 4 - 5 | 679 | 117 | |
XM_006243170.2 | XP_006243232.1 | 4 - 5 | 715 | 117 | ||
GenBank mRNA | BC090343.1 | - | 3 - 4 | 386 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
