Assay Details
Gene Symbol: Adpgk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100723.1 | NP_001094193.1 | 6 - 7 | 979 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P113675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121002 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14237 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P113675 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P113675 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P113675 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P21650 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10910489 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10910486 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10910488 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10910487 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10910484 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10910485 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10910483 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10910482 | - | - |
Rat Genome 230 2.0 Array | 1374138_PM_at | - | - |
Rat Genome 230 2.0 Array | 1374958_PM_at | - | - |
Rat Genome 230 2.0 Array | 1374138_at | - | - |
Rat Genome 230 2.0 Array | 1377952_at | - | - |
Rat Genome 230 2.0 Array | 1395115_PM_at | - | - |
Rat Genome 230 2.0 Array | 1377952_PM_at | - | - |
Rat Genome 230 2.0 Array | 1374958_at | - | - |
Rat Genome 230 2.0 Array | 1395115_at | - | - |
Affymetrix HT Rat Focus Array | 1377952_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1377952_at | - | - |
Affymetrix HT Rat Focus Array | 1374958_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1374958_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0800005802.rn.1 | TC0800000836.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800005801.rn.1 | TC0800000836.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800005799.rn.1 | TC0800000836.rn.1 | RTA TC |
Rat Expression Set 230 | 1374958_at | - | - |
Rat Expression Set 230 | 1374958_PM_at | - | - |
Rat Expression Set 230 | 1377952_PM_at | - | - |
Rat Expression Set 230 | 1377952_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10910482 | - | - |
Rat Genome U34 Set | rc_AI228060_at | - | - |
Rat Genome U34 Set | rc_AA819640_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21427526 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373217 | - | - |
