Assay Details
Gene Symbol: Idh3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053638.1 | NP_446090.1 | 1 - 2 | 86 | 168 | |
GenBank mRNA | AB047541.1 | - | 1 - 2 | 86 | 168 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AB047541.1, NM_053638.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
