Assay Details
Gene Symbol: Hmbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013168.2 | NP_037300.2 | 13 - 14 | 1043 | 85 | |
XM_006242896.1 | XP_006242958.1 | 13 - 14 | 1129 | 85 | ||
XM_006242897.1 | XP_006242959.1 | 12 - 13 | 1051 | 85 | ||
XM_008766148.1 | XP_008764370.1 | 12 - 13 | 1213 | 85 | ||
XM_008766149.1 | XP_008764371.1 | 12 - 13 | 1150 | 85 | ||
GenBank mRNA | BC088162.1 | - | 13 - 14 | 1043 | 7375228 | 85 |
FQ225972.1 | - | 7 - 8 | 528 | 85 | ||
FQ230962.1 | - | 7 - 8 | 528 | 85 | ||
FQ231091.1 | - | 13 - 14 | 1028 | 85 | ||
FQ234793.1 | - | 5 - 6 | 415 | 85 | ||
FQ234870.1 | - | 11 - 12 | 1199 | 85 | ||
X06827.1 | - | 13 - 14 | 918 | 85 | ||
Y12006.1 | - | 13 - 14 | 913 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
