Assay Details
Gene Symbol: Hmbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013168.2 | NP_037300.2 | 1 - 2 | 164 | 160 | |
XM_008766149.1 | XP_008764371.1 | 1 - 2 | 187 | 160 | ||
GenBank mRNA | BC088162.1 | - | 1 - 2 | 164 | 7375228 | 160 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC088162.1, NM_013168.2, XM_008766149.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
