Assay Details
Gene Symbol: Hmbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013168.2 | NP_037300.2 | 2 - 3 | 220 | 112 | |
XM_008766149.1 | XP_008764371.1 | 2 - 3 | 243 | 112 | ||
GenBank mRNA | BC088162.1 | - | 2 - 3 | 220 | 112 | |
Y12006.1 | - | 2 - 3 | 90 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
