Assay Details
Gene Symbol: Abcg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106816.1 | NP_001100286.1 | 1 - 2 | 322 | 86 | |
XM_006242915.2 | XP_006242977.1 | 1 - 2 | 6054 | 86 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001106816.1, XM_006242915.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
