Assay Details
Gene Symbol: Abcg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106816.1 | NP_001100286.1 | 6 - 7 | 1014 | 138 | |
XM_006242915.2 | XP_006242977.1 | 6 - 7 | 6746 | 138 | ||
XM_006242916.2 | XP_006242978.1 | 6 - 7 | 1104 | 138 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
