Assay Details
Gene Symbol: Pif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044253.1 | NP_001037718.1 | 3 - 4 | 807 | 130 | |
XM_006243271.2 | XP_006243333.1 | 3 - 4 | 1744 | 130 | ||
XM_008766324.1 | XP_008764546.1 | 4 - 5 | 1463 | 130 | ||
XM_008766325.1 | XP_008764547.1 | 4 - 5 | 1435 | 130 | ||
XM_008766326.1 | XP_008764548.1 | 4 - 5 | 1394 | 130 | ||
XM_008766327.1 | XP_008764549.1 | 4 - 5 | 1366 | 130 | ||
GenBank mRNA | DQ489562.1 | - | 3 - 4 | 807 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
