Assay Details
Gene Symbol: Pif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044253.1 | NP_001037718.1 | 4 - 5 | 960 | 76 | |
XM_006243271.2 | XP_006243333.1 | 4 - 5 | 1897 | 76 | ||
XM_008766324.1 | XP_008764546.1 | 5 - 6 | 1616 | 76 | ||
XM_008766325.1 | XP_008764547.1 | 5 - 6 | 1588 | 76 | ||
XM_008766326.1 | XP_008764548.1 | 5 - 6 | 1547 | 76 | ||
XM_008766327.1 | XP_008764549.1 | 5 - 6 | 1519 | 76 | ||
GenBank mRNA | DQ489562.1 | - | 4 - 5 | 960 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
