Assay Details
Gene Symbol: Dpagt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199388.1 | NP_955420.1 | 4 - 5 | 775 | 139 | |
XM_006242921.2 | XP_006242983.1 | 4 - 5 | 1126 | 139 | ||
GenBank mRNA | BC063184.1 | - | 4 - 5 | 775 | 139 | |
FQ209511.1 | - | 2 - 3 | 214 | 139 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
