Assay Details
Gene Symbol: Neil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025754.1 | NP_001020925.1 | 1 - 2 | 94 | 63 | |
XM_006243135.2 | XP_006243197.1 | 1 - 2 | 1014 | 63 | ||
GenBank mRNA | BC099125.1 | - | 1 - 2 | 94 | 7445605 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC099125.1, NM_001025754.1, XM_006243135.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
