Assay Details
Gene Symbol: Neil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025754.1 | NP_001020925.1 | 5 - 6 | 830 | 113 | |
XM_006243134.2 | XP_006243196.1 | 5 - 6 | 1644 | 113 | ||
XM_006243135.2 | XP_006243197.1 | 5 - 6 | 1750 | 113 | ||
XM_006243136.2 | XP_006243198.1 | 6 - 7 | 826 | 113 | ||
GenBank mRNA | BC099125.1 | - | 5 - 6 | 830 | 7445605 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
