Assay Details
Gene Symbol: Neil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025754.1 | NP_001020925.1 | 9 - 10 | 1209 | 93 | |
GenBank mRNA | BC099125.1 | - | 9 - 10 | 1209 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
