Assay Details
Gene Symbol: Hexa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004443.1 | NP_001004443.1 | 2 - 3 | 378 | 104 | |
XM_006243162.1 | XP_006243224.1 | 3 - 4 | 248 | 104 | ||
GenBank mRNA | BC082097.1 | - | 2 - 3 | 378 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
