Assay Details
Gene Symbol: Hsdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025697.1 | NP_001020868.1 | 8 - 9 | 981 | 106 | |
XM_008763735.1 | XP_008761957.1 | 8 - 9 | 1024 | 106 | ||
XM_008763736.1 | XP_008761958.1 | 8 - 9 | 1023 | 106 | ||
GenBank mRNA | BC097407.1 | - | 8 - 9 | 981 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
