Assay Details
Gene Symbol: Fsd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763722.1 | XP_008761944.1 | 4 - 5 | 529 | 139 | |
XM_008763723.1 | XP_008761945.1 | 4 - 5 | 528 | 139 | ||
XM_008763724.1 | XP_008761946.1 | 4 - 5 | 529 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() |
