Assay Details
Gene Symbol: Fsd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763722.1 | XP_008761944.1 | 8 - 9 | 983 | 126 | |
XM_008763723.1 | XP_008761945.1 | 8 - 9 | 982 | 126 | ||
XM_008763724.1 | XP_008761946.1 | 8 - 9 | 983 | 126 | ||
XM_008763725.1 | XP_008761947.1 | 7 - 8 | 882 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() |
