Assay Details
Gene Symbol: Rbpjl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108604.1 | NP_001102074.1 | 1 - 2 | 179 | 73 | |
XM_006235598.2 | XP_006235660.1 | 1 - 2 | 259 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108604.1, XM_006235598.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
