Assay Details
Gene Symbol: Rbpjl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108604.1 | NP_001102074.1 | 5 - 6 | 586 | 80 | |
XM_006235598.2 | XP_006235660.1 | 5 - 6 | 666 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
