Assay Details
Gene Symbol: Src | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031977.1 | NP_114183.1 | 2 - 3 | 351 | 94 | |
XM_008762387.1 | XP_008760609.1 | 2 - 3 | 360 | 94 | ||
XM_008762388.1 | XP_008760610.1 | 2 - 3 | 397 | 94 | ||
GenBank mRNA | AF157016.1 | - | 2 - 3 | 351 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
