Assay Details
Gene Symbol: Cyp24a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201635.2 | NP_963966.1 | 4 - 5 | 641 | 92 | |
XM_006235672.2 | XP_006235734.1 | 4 - 5 | 983 | 92 | ||
GenBank mRNA | BC100059.1 | - | 4 - 5 | 986 | 7129117 | 92 |
X59506.1 | - | 4 - 5 | 984 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
