Assay Details
Gene Symbol: Cyp24a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201635.2 | NP_963966.1 | 5 - 6 | 732 | 98 | |
XM_006235672.2 | XP_006235734.1 | 5 - 6 | 1074 | 98 | ||
GenBank mRNA | BC100059.1 | - | 5 - 6 | 1077 | 98 | |
X59506.1 | - | 5 - 6 | 1075 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
