Assay Details
Gene Symbol: Nnat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_073089.1 | 1 - 2 | 194 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
