Assay Details
Gene Symbol: Hnf4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270931.1 | NP_001257860.1 | 9 - 10 | 1304 | 137 | |
NM_001270933.1 | NP_001257862.1 | 9 - 10 | 1304 | 137 | ||
NM_022180.2 | NP_071516.2 | 8 - 9 | 1264 | 137 | ||
XM_006235515.2 | XP_006235577.1 | 8 - 9 | 1259 | 137 | ||
XM_006235516.2 | XP_006235578.1 | 8 - 9 | 1259 | 137 | ||
GenBank mRNA | D10554.1 | - | 8 - 9 | 1140 | 137 | |
FJ608816.1 | - | 9 - 10 | 1304 | 137 | ||
FJ608817.1 | - | 9 - 10 | 1304 | 137 | ||
X57133.1 | - | 8 - 9 | 1155 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
ornithine metabolic process
lipid metabolic process
acyl-CoA metabolic process
xenobiotic metabolic process
establishment of tissue polarity
sex differentiation
blood coagulation
negative regulation of cell proliferation
response to glucose
regulation of gastrulation
regulation of lipid metabolic process
signal transduction involved in regulation of gene expression
cell differentiation
negative regulation of cell growth
intracellular receptor signaling pathway
regulation of microvillus assembly
negative regulation of protein import into nucleus, translocation
response to drug
negative regulation of tyrosine phosphorylation of Stat5 protein
glucose homeostasis
steroid hormone mediated signaling pathway
negative regulation of sequence-specific DNA binding transcription factor activity
cell-cell junction organization
positive regulation of gluconeogenesis
positive regulation of fatty acid biosynthetic process
positive regulation of transcription, DNA-templated
negative regulation of mitotic cell cycle
positive regulation of transcription from RNA polymerase II promoter
regulation of insulin secretion
response to cAMP
lipid homeostasis
phospholipid homeostasis
SMAD protein signal transduction
triglyceride homeostasis
negative regulation of activation of JAK2 kinase activity
positive regulation of cholesterol homeostasis
fatty-acyl-CoA binding
RNA polymerase II core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II activating transcription factor binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
receptor binding
fatty acid binding
protein binding
transcription factor binding
zinc ion binding
palmitoyl-CoA hydrolase activity
protein homodimerization activity
sequence-specific DNA binding
transcription regulatory region DNA binding
repressing transcription factor binding
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
ornithine metabolic process
lipid metabolic process
acyl-CoA metabolic process
xenobiotic metabolic process
establishment of tissue polarity
sex differentiation
blood coagulation
negative regulation of cell proliferation
response to glucose
regulation of gastrulation
regulation of lipid metabolic process
signal transduction involved in regulation of gene expression
cell differentiation
negative regulation of cell growth
intracellular receptor signaling pathway
regulation of microvillus assembly
negative regulation of protein import into nucleus, translocation
response to drug
negative regulation of tyrosine phosphorylation of Stat5 protein
glucose homeostasis
steroid hormone mediated signaling pathway
negative regulation of sequence-specific DNA binding transcription factor activity
cell-cell junction organization
positive regulation of gluconeogenesis
positive regulation of fatty acid biosynthetic process
positive regulation of transcription, DNA-templated
negative regulation of mitotic cell cycle
positive regulation of transcription from RNA polymerase II promoter
regulation of insulin secretion
response to cAMP
lipid homeostasis
phospholipid homeostasis
SMAD protein signal transduction
triglyceride homeostasis
negative regulation of activation of JAK2 kinase activity
positive regulation of cholesterol homeostasis
fatty-acyl-CoA binding
RNA polymerase II core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II activating transcription factor binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
receptor binding
fatty acid binding
protein binding
transcription factor binding
zinc ion binding
palmitoyl-CoA hydrolase activity
protein homodimerization activity
sequence-specific DNA binding
transcription regulatory region DNA binding
repressing transcription factor binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P046573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775030 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297222 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936534 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209221 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88638 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125192 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P046573 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P046573 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P046573 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12146 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P419188 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32392 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3135 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84583 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2664 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 85374 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10842020 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10842019 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10842025 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10842022 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10842021 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10842029 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10842026 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10842027 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10842030 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10842024 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10842023 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10842018 | - | - |
Rat Genome 230 2.0 Array | 1382496_at | - | - |
Rat Genome 230 2.0 Array | 1369289_PM_at | - | - |
Rat Genome 230 2.0 Array | 1382496_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369289_at | - | - |
Affymetrix HT Rat Focus Array | 1369289_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369289_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300016854.rn.1 | TC0300002259.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300016868.rn.1 | TC0300002259.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300016865.rn.1 | TC0300002259.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300016867.rn.1 | TC0300002259.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300016861.rn.1 | TC0300002259.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300016850.rn.1 | TC0300002259.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300016860.rn.1 | TC0300002259.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300016858.rn.1 | TC0300002259.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300016863.rn.1 | TC0300002259.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300016851.rn.1 | TC0300002259.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300016857.rn.1 | TC0300002259.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300016862.rn.1 | TC0300002259.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300016859.rn.1 | TC0300002259.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300016855.rn.1 | TC0300002259.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300016853.rn.1 | TC0300002259.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300016856.rn.1 | TC0300002259.rn.1 | RTA TC |
Rat Expression Set 230 | 1369289_at | - | - |
Rat Expression Set 230 | 1382496_PM_at | - | - |
Rat Expression Set 230 | 1369289_PM_at | - | - |
Rat Expression Set 230 | 1382496_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10842018 | - | - |
Rat Genome U34 Set | X57133mRNA_at | - | - |
Rat Genome U34 Set | D10554_s_at | - | - |
Rat Genome U34 Set | X57133mRNA_g_at | - | - |
Rat Genome U34 Set | rc_AI072326_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1354435 | - | - |
RatRef-12 Expression BeadChip | ILMN_69662 | - | - |
