Assay Details
Gene Symbol: Bcas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145670.4 | NP_663703.3 | 8 - 9 | 1350 | 111 | |
XM_008762466.1 | XP_008760688.1 | 9 - 10 | 1485 | 111 | ||
GenBank mRNA | AB180269.1 | - | 7 - 8 | 993 | 111 | |
BC103480.1 | - | 7 - 8 | 1248 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
