Assay Details
Gene Symbol: Bcas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145670.4 | NP_663703.3 | 1 - 2 | 74 | 64 | |
XM_008762466.1 | XP_008760688.1 | 1 - 2 | 74 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
