Assay Details
Gene Symbol: Pltp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168543.1 | NP_001162015.1 | 1 - 2 | 244 | 78 | |
XM_006235535.2 | XP_006235597.1 | 2 - 3 | 302 | 78 | ||
XM_008762476.1 | XP_008760698.1 | 2 - 3 | 296 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001168543.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
