Assay Details
Gene Symbol: Pltp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168543.1 | NP_001162015.1 | 4 - 5 | 585 | 68 | |
XM_006235535.2 | XP_006235597.1 | 5 - 6 | 643 | 68 | ||
XM_008762476.1 | XP_008760698.1 | 5 - 6 | 637 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
