Assay Details
Gene Symbol: Cd40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134360.1 | NP_599187.1 | 1 - 2 | 79 | 91 | |
XM_006235511.2 | XP_006235573.1 | 1 - 2 | 141 | 91 | ||
XM_008762463.1 | XP_008760685.1 | 1 - 2 | 142 | 91 | ||
XM_008762464.1 | XP_008760686.1 | 1 - 2 | 140 | 91 | ||
XM_008762465.1 | XP_008760687.1 | 1 - 2 | 141 | 91 | ||
GenBank mRNA | AF241231.2 | - | 1 - 2 | 94 | 91 | |
BC097949.1 | - | 1 - 2 | 79 | 7387440 | 91 | |
FQ226992.1 | - | 1 - 2 | 62 | 91 | ||
FQ231420.1 | - | 1 - 2 | 82 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF241231.2, BC097949.1, NM_134360.1, XM_006235511.2, XM_008762463.1, XM_008762464.1, XM_008762465.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
