Assay Details
Gene Symbol: Cd40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134360.1 | NP_599187.1 | 5 - 6 | 520 | 129 | |
XM_006235511.2 | XP_006235573.1 | 5 - 6 | 582 | 129 | ||
XM_008762464.1 | XP_008760686.1 | 5 - 6 | 581 | 129 | ||
GenBank mRNA | BC097949.1 | - | 5 - 6 | 520 | 129 | |
FQ226992.1 | - | 5 - 6 | 503 | 129 | ||
FQ231420.1 | - | 5 - 6 | 523 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
