Assay Details
Gene Symbol: Ptpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012637.2 | NP_036769.1 | 1 - 2 | 135 | 143 | |
XM_006235638.2 | XP_006235700.1 | 1 - 2 | 204 | 143 | ||
XM_006235639.2 | XP_006235701.1 | 1 - 2 | 205 | 143 | ||
GenBank mRNA | BC081829.1 | - | 1 - 2 | 135 | 7135855 | 143 |
M33962.1 | - | 1 - 2 | 177 | 143 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC081829.1, M33962.1, NM_012637.2, XM_006235638.2, XM_006235639.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
