Assay Details
Gene Symbol: Tnnc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037351.1 | NP_001032428.1 | 2 - 3 | 201 | 80 | |
GenBank mRNA | DQ273679.1 | - | 2 - 3 | 201 | 80 | |
FQ215069.1 | - | 265 | 80 | |||
FQ216869.1 | - | 265 | 80 | |||
FQ216875.1 | - | 295 | 80 | |||
FQ216940.1 | - | 257 | 80 | |||
FQ217047.1 | - | 265 | 80 | |||
FQ217054.1 | - | 80 | ||||
FQ217106.1 | - | 294 | 80 | |||
FQ217137.1 | - | 265 | 80 | |||
FQ217169.1 | - | 265 | 80 | |||
FQ217172.1 | - | 265 | 80 | |||
FQ217236.1 | - | 276 | 80 | |||
FQ217299.1 | - | 265 | 80 | |||
FQ217371.1 | - | 265 | 80 | |||
FQ217378.1 | - | 265 | 80 | |||
FQ217706.1 | - | 265 | 80 | |||
FQ217788.1 | - | 265 | 80 | |||
FQ217840.1 | - | 265 | 80 | |||
FQ217842.1 | - | 265 | 80 | |||
FQ217896.1 | - | 264 | 80 | |||
FQ217902.1 | - | 265 | 80 | |||
FQ217984.1 | - | 265 | 80 | |||
FQ218010.1 | - | 265 | 80 | |||
FQ218048.1 | - | 295 | 80 | |||
FQ218061.1 | - | 265 | 80 | |||
FQ223543.1 | - | 265 | 80 | |||
FQ223572.1 | - | 265 | 80 | |||
FQ223583.1 | - | 265 | 80 | |||
FQ223607.1 | - | 265 | 80 | |||
FQ223623.1 | - | 265 | 80 | |||
FQ223636.1 | - | 265 | 80 | |||
FQ223643.1 | - | 265 | 80 | |||
FQ223690.1 | - | 265 | 80 | |||
FQ223692.1 | - | 265 | 80 | |||
FQ223711.1 | - | 265 | 80 | |||
FQ223713.1 | - | 265 | 80 | |||
FQ223719.1 | - | 265 | 80 | |||
FQ223773.1 | - | 277 | 80 | |||
FQ223816.1 | - | 286 | 80 | |||
FQ223825.1 | - | 265 | 80 | |||
FQ223832.1 | - | 265 | 80 | |||
FQ223835.1 | - | 295 | 80 | |||
FQ223976.1 | - | 265 | 80 | |||
FQ223986.1 | - | 265 | 80 | |||
FQ224079.1 | - | 265 | 80 | |||
FQ224114.1 | - | 294 | 80 | |||
FQ224128.1 | - | 265 | 80 | |||
FQ224223.1 | - | 265 | 80 | |||
FQ224258.1 | - | 265 | 80 | |||
FQ224340.1 | - | 265 | 80 | |||
FQ224376.1 | - | 280 | 80 | |||
FQ224378.1 | - | 265 | 80 | |||
FQ224450.1 | - | 265 | 80 | |||
FQ224462.1 | - | 265 | 80 | |||
FQ224599.1 | - | 265 | 80 | |||
FQ224648.1 | - | 265 | 80 | |||
FQ224672.1 | - | 265 | 80 | |||
FQ224735.1 | - | 266 | 80 | |||
FQ224749.1 | - | 265 | 80 | |||
FQ224755.1 | - | 80 | ||||
FQ224795.1 | - | 265 | 80 | |||
FQ224811.1 | - | 265 | 80 | |||
FQ224822.1 | - | 265 | 80 | |||
FQ224827.1 | - | 262 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
