Assay Details
Gene Symbol: Tnnc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FQ215069.1 | - | 519 | 76 | ||
FQ216531.1 | - | 519 | 76 | |||
FQ216869.1 | - | 519 | 76 | |||
FQ216875.1 | - | 549 | 76 | |||
FQ216940.1 | - | 511 | 76 | |||
FQ216956.1 | - | 76 | ||||
FQ217047.1 | - | 519 | 76 | |||
FQ217054.1 | - | 519 | 76 | |||
FQ217106.1 | - | 548 | 76 | |||
FQ217137.1 | - | 519 | 76 | |||
FQ217169.1 | - | 519 | 76 | |||
FQ217195.1 | - | 519 | 76 | |||
FQ217236.1 | - | 530 | 76 | |||
FQ217299.1 | - | 519 | 76 | |||
FQ217371.1 | - | 519 | 76 | |||
FQ217378.1 | - | 519 | 76 | |||
FQ217706.1 | - | 519 | 76 | |||
FQ217788.1 | - | 519 | 76 | |||
FQ217840.1 | - | 519 | 76 | |||
FQ217842.1 | - | 519 | 76 | |||
FQ217896.1 | - | 518 | 76 | |||
FQ217974.1 | - | 548 | 76 | |||
FQ217984.1 | - | 519 | 76 | |||
FQ218010.1 | - | 519 | 76 | |||
FQ218048.1 | - | 549 | 76 | |||
FQ223543.1 | - | 519 | 76 | |||
FQ223572.1 | - | 519 | 76 | |||
FQ223583.1 | - | 519 | 76 | |||
FQ223607.1 | - | 519 | 76 | |||
FQ223623.1 | - | 519 | 76 | |||
FQ223636.1 | - | 519 | 76 | |||
FQ223643.1 | - | 519 | 76 | |||
FQ223690.1 | - | 519 | 76 | |||
FQ223692.1 | - | 519 | 76 | |||
FQ223711.1 | - | 519 | 76 | |||
FQ223713.1 | - | 519 | 76 | |||
FQ223719.1 | - | 519 | 76 | |||
FQ223773.1 | - | 531 | 76 | |||
FQ223816.1 | - | 540 | 76 | |||
FQ223825.1 | - | 519 | 76 | |||
FQ223835.1 | - | 549 | 76 | |||
FQ223976.1 | - | 519 | 76 | |||
FQ223986.1 | - | 519 | 76 | |||
FQ224114.1 | - | 548 | 76 | |||
FQ224128.1 | - | 519 | 76 | |||
FQ224223.1 | - | 519 | 76 | |||
FQ224258.1 | - | 519 | 76 | |||
FQ224340.1 | - | 519 | 76 | |||
FQ224376.1 | - | 534 | 76 | |||
FQ224378.1 | - | 519 | 76 | |||
FQ224387.1 | - | 519 | 76 | |||
FQ224450.1 | - | 519 | 76 | |||
FQ224599.1 | - | 519 | 76 | |||
FQ224648.1 | - | 519 | 76 | |||
FQ224749.1 | - | 519 | 76 | |||
FQ224755.1 | - | 548 | 76 | |||
FQ224811.1 | - | 519 | 76 | |||
FQ224822.1 | - | 519 | 76 | |||
FQ224827.1 | - | 516 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
