Assay Details
Gene Symbol: Tomm34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044244.2 | NP_001037709.1 | 1 - 2 | 145 | 90 | |
GenBank mRNA | BC105768.1 | - | 1 - 2 | 146 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
