Assay Details
Gene Symbol: Tomm34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044244.2 | NP_001037709.1 | 2 - 3 | 244 | 88 | |
XM_006235585.2 | XP_006235647.1 | 2 - 3 | 519 | 88 | ||
GenBank mRNA | BC105768.1 | - | 2 - 3 | 245 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
