Assay Details
Gene Symbol: Tomm34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044244.2 | NP_001037709.1 | 4 - 5 | 564 | 72 | |
XM_006235585.2 | XP_006235647.1 | 4 - 5 | 839 | 72 | ||
GenBank mRNA | BC105768.1 | - | 4 - 5 | 565 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
