Assay Details
Gene Symbol: Pkig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153469.4 | NP_703199.1 | 1 - 2 | 44 | 66 | |
GenBank mRNA | BC088108.1 | - | 1 - 2 | 44 | 66 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC088108.1, NM_153469.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
