Assay Details
Gene Symbol: Pkig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006235518.2 | XP_006235580.1 | 1 - 2 | 1291 | 83 | |
GenBank mRNA | FQ218619.1 | - | 1 - 2 | 146 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
