Assay Details
Gene Symbol: Gk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024381.2 | NP_077357.2 | 13 - 14 | 1136 | 106 | |
XM_006256987.2 | XP_006257049.1 | 14 - 15 | 1188 | 106 | ||
XM_006256988.2 | XP_006257050.1 | 13 - 14 | 1170 | 106 | ||
XM_008773274.1 | XP_008771496.1 | 15 - 16 | 1253 | 106 | ||
XM_008773275.1 | XP_008771497.1 | 14 - 15 | 1235 | 106 | ||
XM_008773276.1 | XP_008771498.1 | 15 - 16 | 1253 | 106 | ||
XM_008773278.1 | XP_008771500.1 | 15 - 16 | 1252 | 106 | ||
GenBank mRNA | CO564445.1 | - | 4 - 5 | 269 | 106 | |
D16102.1 | - | 13 - 14 | 1139 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
