Assay Details
Gene Symbol: Pdk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106581.1 | NP_001100051.1 | 10 - 11 | 1146 | 63 | |
XM_006257018.2 | XP_006257080.1 | 9 - 10 | 1213 | 63 | ||
XM_006257019.2 | XP_006257081.1 | 9 - 10 | 1224 | 63 | ||
XM_006257020.2 | XP_006257082.1 | 9 - 10 | 1215 | 63 | ||
GenBank mRNA | BC169078.1 | - | 9 - 10 | 1160 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
