Assay Details
Gene Symbol: Pdk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106581.1 | NP_001100051.1 | 12 - 13 | 1391 | 86 | |
XM_006257018.2 | XP_006257080.1 | 11 - 12 | 1458 | 86 | ||
XM_006257019.2 | XP_006257081.1 | 11 - 12 | 1469 | 86 | ||
XM_006257020.2 | XP_006257082.1 | 11 - 12 | 1460 | 86 | ||
GenBank mRNA | BC169078.1 | - | 11 - 12 | 1405 | 5598223 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
regulation of acetyl-CoA biosynthetic process from pyruvate
regulation of glucose metabolic process
peptidyl-serine phosphorylation
peroxisome proliferator activated receptor signaling pathway
cellular response to glucose stimulus
cellular response to fatty acid
hypoxia-inducible factor-1alpha signaling pathway
regulation of reactive oxygen species metabolic process
molecular_function
protein kinase activity
protein serine/threonine kinase activity
pyruvate dehydrogenase (acetyl-transferring) kinase activity
ATP binding
biological_process
regulation of acetyl-CoA biosynthetic process from pyruvate
regulation of glucose metabolic process
peptidyl-serine phosphorylation
peroxisome proliferator activated receptor signaling pathway
cellular response to glucose stimulus
cellular response to fatty acid
hypoxia-inducible factor-1alpha signaling pathway
regulation of reactive oxygen species metabolic process
molecular_function
protein kinase activity
protein serine/threonine kinase activity
pyruvate dehydrogenase (acetyl-transferring) kinase activity
ATP binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P991682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829985 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743739 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149745 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P991682 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P991682 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P991682 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P443343 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P991682 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32064 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 51818 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10938389 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938385 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938394 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938386 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938390 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938392 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938387 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938393 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938388 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938391 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10938384 | - | - |
Rat Genome 230 2.0 Array | 1372229_at | - | - |
Rat Genome 230 2.0 Array | 1384315_at | - | - |
Rat Genome 230 2.0 Array | 1372229_PM_at | - | - |
Rat Genome 230 2.0 Array | 1384315_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1384315_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1384315_at | - | - |
Affymetrix HT Rat Focus Array | 1372229_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372229_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0X00011734.rn.1 | TC0X00001861.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00011736.rn.1 | TC0X00001861.rn.1 | RTA TC |
Rat Expression Set 230 | 1372229_PM_at | - | - |
Rat Expression Set 230 | 1372229_at | - | - |
Rat Expression Set 230 | 1384315_at | - | - |
Rat Expression Set 230 | 1384315_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938384 | - | - |
Rat Genome U34 Set | rc_AI231465_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20714771 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365682 | - | - |
