Assay Details
Gene Symbol: Pdk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106581.1 | NP_001100051.1 | 3 - 4 | 430 | 94 | |
XM_006257018.2 | XP_006257080.1 | 2 - 3 | 497 | 94 | ||
XM_006257019.2 | XP_006257081.1 | 2 - 3 | 508 | 94 | ||
XM_006257020.2 | XP_006257082.1 | 2 - 3 | 499 | 94 | ||
GenBank mRNA | BC169078.1 | - | 2 - 3 | 444 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
