Assay Details
Gene Symbol: Atf7ip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006245728.2 | XP_006245790.1 | 11 | 2297 | 195 | |
XM_006245732.2 | XP_006245794.1 | 11 | 2263 | 195 | ||
XM_006245735.1 | XP_006245797.1 | 9 | 1871 | 195 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P116774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672471 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752054 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878033 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42629 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P116774 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P116774 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P116774 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P173610 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 63892 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 9075 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95136 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10740490 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740495 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740480 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740485 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740487 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740489 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740494 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740482 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740488 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740486 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740491 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740481 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10740479 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740479 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21783824 | - | - |
ABI Rat Genome Survey Microarray | 22187687 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355887 | - | - |
