Assay Details
Gene Symbol: Rbfox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106974.2 | NP_001100444.2 | 1 | 240 | 80 | |
XM_006245774.2 | XP_006245836.1 | 1 | 774 | 80 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001106974.2, XM_006245774.2 transcripts.
This assay is in the 5'UTR region of the NM_001106974.2, XM_006245774.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
