Assay Details
Gene Symbol: Dnase1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013097.2 | NP_037229.1 | 1 - 2 | 278 | 72 | |
XM_006245829.2 | XP_006245891.1 | 2 - 3 | 958 | 72 | ||
GenBank mRNA | AF397150.1 | - | 1 - 2 | 278 | 72 | |
AF397151.1 | - | 2 - 3 | 149 | 72 | ||
BC089764.1 | - | 1 - 2 | 278 | 7130586 | 72 | |
X56060.1 | - | 1 - 2 | 131 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF397150.1, AF397151.1, BC089764.1, NM_013097.2, XM_006245829.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
